N-(tert-Butoxycarbonyl)-2-(2-aminoethoxy)ethylamine ≥95.0% (by GC)

Supplier: TCI America

Synonyms: tert-Butyl (2-(2-aminoethoxy)ethyl)carbamate, N-Boc-2-(2-aminoethoxy)ethylamine, Amino-PEG2-NH-Boc, [2-(2-Aminoethoxy)ethyl]carbamic acid tert-Butyl ester, tert-Butyl [2-(2-Aminoethoxy)ethyl]carbamate

B5683-1G B5683-200MG
TCB5683-1GEA 93 USD
TCB5683-1G TCB5683-200MG
N-(tert-Butoxycarbonyl)-2-(2-aminoethoxy)ethylamine ≥95.0% (by GC)
N-(tert-Butoxycarbonyl)-2-(2-aminoethoxy)ethylamine
Formula: C₉H₂₀N₂O₃
MW: 204.27 g/mol
MDL Number: MFCD12031501
CAS Number: 127828-22-2

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR