4-Chloro-N-methoxy-N-methylbenzamide ≥98.0% (by GC)

Supplier: TCI America
Warning

C3486-25G C3486-5G
TCC3486-25GEA 349 USD
TCC3486-25G TCC3486-5G
4-Chloro-N-methoxy-N-methylbenzamide ≥98.0% (by GC)
4-Chloro-N-methoxy-N-methylbenzamide
Formula: C₉H₁₀ClNO₂
MDL Number: MFCD02684308
CAS Number: 122334-37-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR