Adamantan-1-yl acrylate ≥98.0% (by GC) stabilised

Supplier: TCI America
Warning

Synonyms: Acrylic acid Adamantan-1-yl ester, 1-Acryloyloxyadamantane

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A1735-1G A1735-5G
TCA1735-1GEA 97 USD
TCA1735-1G TCA1735-5G
Adamantan-1-yl acrylate ≥98.0% (by GC) stabilised
Adamantan-1-yl acrylate
Formula: C₁₃H₁₈O₂
MDL Number: MFCD18072700
CAS Number: 121601-93-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR