(7-Bromo-9,9-dimethyl-9H-fluoren-2-yl)boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

B5773-1G B5773-5G
TCB5773-1GEA 120 USD
TCB5773-1G TCB5773-5G
(7-Bromo-9,9-dimethyl-9H-fluoren-2-yl)boronic acid (contains varying amounts of Anhydride)
(7-Bromo-9,9-dimethyl-9H-fluoren-2-yl)boronic acid
Formula: C₁₅H₁₄BBrO₂
MDL Number: MFCD26403351
CAS Number: 1213768-48-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 98.0 to 105.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR