Biapenem ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (4R,5S,6S)-3-((6,7-Dihydro-5H-pyrazolo[1,2-a][1,2,4]triazol-4-ium-6-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate

B5522-100MG B5522-25MG
TCB5522-100MGEA 314 USD
TCB5522-100MG TCB5522-25MG
Biapenem ≥98.0% (by HPLC)
Biapenem
Formula: C₁₅H₁₈N₄O₄S
MW: 350.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00864863
CAS Number: 120410-24-4
Merck Index: 14,01197

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 15.20 to 16.70 %
Specific rotation [a]20/D -30.0 to 335.0 deg (C=0.5, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR