(3-Bromophenyl)(mesityl)iodonium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: (3-Bromophenyl)(mesityl)iodonium triflate, (3-Bromophenyl)(2,4,6-trimethylphenyl)iodonium triflate

B5277-1G B5277-5G
TCB5277-1GEA 137 USD
TCB5277-1G TCB5277-5G
(3-Bromophenyl)(mesityl)iodonium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
(3-Bromophenyl)(mesityl)iodonium trifluoromethanesulfonate
Formula: C₁₆H₁₅BrF₃IO₃S
MW: 551.16 g/mol
MDL Number: MFCD20264881
CAS Number: 1203709-76-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Ion exchange titration) min. 98.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR