N,N'-Bis[3-(dimethylamino)propyl]perylene-3,4,9,10-tetracarboxylic diimide ≥95.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
DAPER, 2,9-Bis[3-(dimethylamino)propyl]anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetrone
B5954-1G
B5954-200MG
TCB5954-1GEA
428
USD
TCB5954-1G
TCB5954-200MG
N,N'-Bis[3-(dimethylamino)propyl]perylene-3,4,9,10-tetracarboxylic diimide ≥95.0% (by total nitrogen basis)
N,N'-Bis[3-(dimethylamino)propyl]perylene-3,4,9,10-tetracarboxylic diimide
Formula:
C₃₄H₃₂N₄O₄ MW: 560.65 g/mol Melting Pt: 300 °C |
MDL Number:
MFCD03791075 CAS Number: 117901-97-0 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (with Total Nitrogen) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...