4-(3-Buten-1-yl)-4'-methylbiphenyl ≥98.0% (by GC)

Supplier: TCI America

B5872-1G B5872-5G
TCB5872-1GEA 93 USD
TCB5872-1G TCB5872-5G
4-(3-Buten-1-yl)-4'-methylbiphenyl ≥98.0% (by GC)
4-(3-Buten-1-yl)-4'-methylbiphenyl
Formula: C₁₇H₁₈
MW: 222.33 g/mol
CAS Number: 117713-14-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR