2-Bromoindolo[3,2,1-jk]carbazole ≥98.0% (by GC)

Supplier: TCI America

B5397-1G B5397-200MG
TCB5397-1GEA 301 USD
TCB5397-1G TCB5397-200MG
2-Bromoindolo[3,2,1-jk]carbazole ≥98.0% (by GC)
2-Bromoindolo[3,2,1-jk]carbazole
Formula: C₁₈H₁₀BrN
MW: 320.19 g/mol
MDL Number: MFCD30187281
CAS Number: 1174032-81-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 216.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR