Benzyl (6-Bromohexyl)carbamate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 6-[N-(Carbobenzoxy)amino]hexyl bromide, Benzyl (6-bromohexyl)carbamate, 6-(N-Benzyloxycarbonylamino)hexyl bromide, 6-(N-Cbz-amino)hexyl bromide, (6-Bromohexyl)carbamic acid Benzyl ester

B5355-1G B5355-200MG
TCB5355-1GEA 214 USD
TCB5355-1G TCB5355-200MG
Benzyl (6-Bromohexyl)carbamate ≥98.0% (by HPLC, total nitrogen)
Benzyl (6-Bromohexyl)carbamate
Formula: C₁₄H₂₀BrNO₂
MW: 314.22 g/mol
MDL Number: MFCD08436190
CAS Number: 116784-97-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 49.0 to 53.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR