3-Bromo-2-methoxyaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

B5131-1G B5131-5G
TCB5131-1GEA 99 USD
TCB5131-1G TCB5131-5G
3-Bromo-2-methoxyaniline ≥98.0% (by GC, titration analysis)
3-Bromo-2-methoxyaniline
Formula: C₇H₈BrNO
Boiling Pt: 159 °C
MDL Number: MFCD13186737
CAS Number: 116557-46-1
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR