Bis(3-bromophenyl)phenylphosphine oxide ≥98.0% (by GC)

Supplier: TCI America
Warning

B5531-1G B5531-200MG
TCB5531-1GEA 223 USD
TCB5531-1G TCB5531-200MG
Bis(3-bromophenyl)phenylphosphine oxide ≥98.0% (by GC)
Bis(3-bromophenyl)phenylphosphine oxide
Formula: C₁₈H₁₃Br₂OP
MW: 436.08 g/mol
CAS Number: 1163698-32-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 112.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR