4,8-Bis[(2-ethylhexyl)oxy]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene ≥97.0% (by HPLC)
Supplier: TCI America
|
Danger
|
Synonyms:
[4,8-Bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b']dithiophene-2,6-diyl]bis(trimethylstannane), (4,8-Bis((2-ethylhexyl)oxy)benzo[1,2-b:4,5-b']dithiophene-2,6-diyl)bis(trimethylstannane)
B5490-500MG
B5490-5G
TCB5490-500MGEA
125
USD
TCB5490-500MG
TCB5490-5G
4,8-Bis[(2-ethylhexyl)oxy]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene ≥97.0% (by HPLC)
4,8-Bis[(2-ethylhexyl)oxy]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene
Formula:
C₃₂H₅₄O₂S₂Sn₂ MW: 772.32 g/mol |
MDL Number:
MFCD20275089 CAS Number: 1160823-78-8 UN: 3146 ADR: 6.1,I |
Specification Test Results
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Sulfuride) | 7.80 to 8.80 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...