1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine ≥95.0% (by GC)

Supplier: TCI America
Danger

Synonyms: tert-Butyl 3-(Cyanomethylene)azetidine-1-carboxylate, 3-(Cyanomethylene)azetidine-1-carboxylic acid tert-Butyl ester, 1-Boc-3-(cyanomethylene)azetidine

B5217-1G B5217-200MG
TCB5217-1GEA 134 USD
TCB5217-1G TCB5217-200MG
1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine ≥95.0% (by GC)
1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine
Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12761241
CAS Number: 1153949-11-1
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR