Chlorprothixene ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: (Z)-2-Chloro-9-(3-dimethylaminopropylidene)thioxanthene, (Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)-N,N-dimethyl-1-propanamine

C3505-1G C3505-250MG C3505-5G
TCC3505-1GEA 188 USD
TCC3505-1G TCC3505-250MG TCC3505-5G
Chlorprothixene ≥98.0% (by titrimetric analysis)
Chlorprothixene
Formula: C₁₈H₁₈ClNS
MW: 315.86 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00869180
CAS Number: 113-59-7
EINECS: 204-032-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,02188

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR