Bis(1,10-phenanthroline)palladium(II) bis(hexafluorophosphate) ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B5400-1G B5400-200MG
TCB5400-1GEA 395 USD
TCB5400-1G TCB5400-200MG
Bis(1,10-phenanthroline)palladium(II) bis(hexafluorophosphate) ≥98.0% (by titrimetric analysis)
Bis(1,10-phenanthroline)palladium(II) bis(hexafluorophosphate)
Formula: C₂₄H₁₆F₁₂N₄P₂Pd
MW: 756.77 g/mol
CAS Number: 113173-22-1

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR