N-(tert-Butoxycarbonyl)-DL-tryptophan ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B4891-1G B4891-5G
TCB4891-1GEA 24 USD
TCB4891-1G TCB4891-5G
N-(tert-Butoxycarbonyl)-DL-tryptophan ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-DL-tryptophan
Formula: C₁₆H₂₀N₂O₄
MDL Number: MFCD00190836
CAS Number: 112525-72-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR