4-Bromo-9-phenyl-9H-carbazole ≥97.0% (by GC)

Supplier: TCI America
Warning

B5767-1G B5767-200MG
TCB5767-1GEA 247 USD
TCB5767-1G TCB5767-200MG
4-Bromo-9-phenyl-9H-carbazole ≥97.0% (by GC)
4-Bromo-9-phenyl-9H-carbazole
Formula: C₁₈H₁₂BrN
MDL Number: MFCD29067336
CAS Number: 1097884-37-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR