Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethyl-3,5-dimethoxybenzylamine)palladium(II)] ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: SingaCycle(TM)-A2

C2406-1G C2406-200MG
TCC2406-1GEA 214 USD
TCC2406-1G TCC2406-200MG
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethyl-3,5-dimethoxybenzylamine)palladium(II)] ≥98.0% (by titrimetric analysis)
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethyl-3,5-dimethoxybenzylamine)palladium(II)]

This product is unavailable in the U.S.

Formula: C₃₈H₅₂ClN₃O₂Pd
MW: 724.72 g/mol
CAS Number: 1093348-08-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR