Dichlorobis(2-ethylhexyl)silane ≥95.0% (by GC)

Supplier: TCI America
Danger

D4443-1G D4443-5G
TCD4443-1GEA 96 USD
TCD4443-1G TCD4443-5G
Dichlorobis(2-ethylhexyl)silane ≥95.0% (by GC)
Dichlorobis(2-ethylhexyl)silane
Formula: C₁₆H₃₄Cl₂Si
MDL Number: MFCD22370170
CAS Number: 1089687-03-5
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Orange to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR