Benzyl N,N,N',N'-Tetraisopropylphosphorodiamidite ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-(Benzyloxy)-N,N,N',N'-tetraisopropylphosphinediamine

B5997-1G B5997-5G
TCB5997-1GEA 151 USD
TCB5997-1G TCB5997-5G
Benzyl N,N,N',N'-Tetraisopropylphosphorodiamidite ≥98.0% (by GC, titration analysis)
Benzyl N,N,N',N'-Tetraisopropylphosphorodiamidite

Benzyl N,N,N',N'-tetraisopropylphosphorodiamidite [B5997] is used in the synthesis of phosphodiesters and phosphorylation of inositols and nucleosides.


B5997 promptly reacts with alcohols in the presence of a base like 1H-tetrazole to afford a phosphoramidite. It can further react with another alcohol resulting in a phosphite triester with different alcohol species. Successive oxidation of the phosphorus atom and deprotection gives a phosphodiester.

Formula: C₁₉H₃₅N₂OP
Storage Temperature: Refrigerator
MDL Number: MFCD32062870
CAS Number: 108549-21-9

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR