tert-Butyl (S)-4-(Hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylate ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (S)-3-(tert-Butoxycarbonyl)-4-(hydroxymethyl)-2,2-dimethyl-1,3-oxazolidine, (S)-3-Boc-4-(hydroxymethyl)-2,2-dimethyl-1,3-oxazolidine, (S)-4-(Hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylic acid tert-Butyl ester

B5657-1G B5657-5G
TCB5657-1GEA 143 USD
TCB5657-1G TCB5657-5G
tert-Butyl (S)-4-(Hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylate ≥97.0% (by GC)
tert-Butyl (S)-4-(Hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylate
Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11041410
CAS Number: 108149-65-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Specific rotation [a]20/D +22.0 to +26.0 deg (C=2, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR