Chloro[(1,3-dimesitylimidazol-2-ylidene)(N,N-dimethylbenzylamine)palladium(II)] ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: SingaCycle(TM)-E1

C2347-1G C2347-200MG C2347-5G
TCC2347-1GEA 261 USD
TCC2347-1G TCC2347-200MG TCC2347-5G
Chloro[(1,3-dimesitylimidazol-2-ylidene)(N,N-dimethylbenzylamine)palladium(II)] ≥98.0% (by titrimetric analysis)
Chloro[(1,3-dimesitylimidazol-2-ylidene)(N,N-dimethylbenzylamine)palladium(II)]

This product is unavailable in the U.S.

Formula: C₃₀H₃₆ClN₃Pd
MW: 580.51 g/mol
CAS Number: 1058661-78-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Purity (Chelometric back titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR