Cerium(III) bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Bis(trifluoromethanesulphonyl)imide Cerium(III) salt, Cerium(III) triflimide

C3516-1G C3516-5G
TCC3516-1GEA 90 USD
TCC3516-1G TCC3516-5G
Cerium(III) bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
Cerium(III) bis(trifluoromethanesulfonyl)imide
Formula: C₆CeF₁₈N₃O₁₂S₆
MW: 980.52 g/mol
CAS Number: 1046099-39-1
UN: 1759
ADR: 8 II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR