9-(4-Bromobutyl)-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5547-1G B5547-200MG
TCB5547-1GEA 204 USD
TCB5547-1G TCB5547-200MG
9-(4-Bromobutyl)-9H-carbazole ≥98.0% (by GC)
9-(4-Bromobutyl)-9H-carbazole
Formula: C₁₆H₁₆BrN
MDL Number: MFCD11546053
CAS Number: 10420-20-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR