Acetamidine hydrobromide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
A3292-1G
A3292-5G
TCA3292-1GEA
52
USD
TCA3292-1G
TCA3292-5G
Acetamidine hydrobromide ≥98.0% (by titrimetric analysis)
Acetamidine hydrobromide
Formula:
C₂H₆N₂·HBr |
CAS Number:
1040352-82-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...