3'-Bromo[1,1'-biphenyl]-3-carbonitrile ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5964-1G
B5964-5G
TCB5964-1GEA
113
USD
TCB5964-1G
TCB5964-5G
3'-Bromo[1,1'-biphenyl]-3-carbonitrile ≥98.0% (by GC)
3'-Bromo[1,1'-biphenyl]-3-carbonitrile
Formula:
C₁₃H₈BrN |
CAS Number:
1040141-34-1 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...