O-(tert-Butyldiphenylsilyl)hydroxylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5999-1G B5999-200MG
TCB5999-1GEA 129 USD
TCB5999-1G TCB5999-200MG
O-(tert-Butyldiphenylsilyl)hydroxylamine ≥98.0% (by GC)
O-(tert-Butyldiphenylsilyl)hydroxylamine
Formula: C₁₆H₂₁NOSi
MDL Number: MFCD06809831
CAS Number: 103587-51-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR