1-Benzylpiperidine-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B5826-1G
B5826-5G
TCB5826-1GEA
132
USD
TCB5826-1G
TCB5826-5G
1-Benzylpiperidine-4-carboxylic acid ≥98.0% (by GC, titration analysis)
1-Benzylpiperidine-4-carboxylic acid
Formula:
C₁₃H₁₇NO₂ MW: 219.28 g/mol |
MDL Number:
MFCD03371463 CAS Number: 10315-07-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...