2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene ≥98.0% (by GC)

Supplier: TCI America

B5335-1G B5335-200MG
TCB5335-1GEA 89 USD
TCB5335-1G TCB5335-200MG
2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene ≥98.0% (by GC)
2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene
Formula: C₁₈H₁₄BrFS
MW: 361.27 g/mol
MDL Number: MFCD21496340
CAS Number: 1030825-20-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C
melting range max. 5.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR