Adamantane-1,3-diamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

A3265-1G A3265-5G
TCA3265-1GEA 135 USD
TCA3265-1G TCA3265-5G
Adamantane-1,3-diamine ≥98.0% (by GC, titration analysis)
Adamantane-1,3-diamine
Formula: C₁₀H₁₈N₂
MDL Number: MFCD01831403
CAS Number: 10303-95-4
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR