2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-[[tert-Butyl(dimethyl)silyl]oxy]ethan-1-ol, 2-((tert-Butyldimethylsilyl)oxy)ethanol

B5851-100G B5851-25G
TCB5851-100GEA 335 USD
TCB5851-100G TCB5851-25G
2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol ≥98.0% (by GC)
2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol
Formula: C₈H₂₀O₂Si
MW: 176.33 g/mol
MDL Number: MFCD09261150
CAS Number: 102229-10-7

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR