1-[4-(4-Amino-3-methoxyphenyl)piperazin-1-yl]ethanone ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 1-(4-(4-Amino-3-methoxyphenyl)piperazin-1-yl)ethanone

A3255-1G A3255-5G
TCA3255-1GEA 34 USD
TCA3255-1G TCA3255-5G
1-[4-(4-Amino-3-methoxyphenyl)piperazin-1-yl]ethanone ≥98.0% (by HPLC)
1-[4-(4-Amino-3-methoxyphenyl)piperazin-1-yl]ethanone
Formula: C₁₃H₁₉N₃O₂
MW: 249.31 g/mol
MDL Number: MFCD19174778
CAS Number: 1021426-42-5

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 129.0 to 133.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR