6,6''-Dibromo-2,2':6',2''-terpyridine ≥94.0% (by GC)

Supplier: TCI America
Warning

D5432-1G D5432-200MG
TCD5432-1GEA 452 USD
TCD5432-1G TCD5432-200MG
6,6''-Dibromo-2,2':6',2''-terpyridine ≥94.0% (by GC)
6,6''-Dibromo-2,2':6',2''-terpyridine
Formula: C₁₅H₉Br₂N₃
MW: 391.07 g/mol
MDL Number: MFCD01863557
CAS Number: 100366-66-3

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR