4,4'-Dibromo-4''-methoxytriphenylamine ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 4-Bromo-N-(4-bromophenyl)-N-(4-methoxyphenyl)aniline, N,N-Bis(4-bromophenyl)-4-methoxyaniline

D5339-1G D5339-5G
TCD5339-1GEA 117 USD
TCD5339-1G TCD5339-5G
4,4'-Dibromo-4''-methoxytriphenylamine ≥98.0% (by total nitrogen basis)
4,4'-Dibromo-4''-methoxytriphenylamine
Formula: C₁₉H₁₅Br₂NO
MDL Number: MFCD31699798
CAS Number: 100308-69-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 73.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR