2-Bromo-5-tert-butyl-1,3-dimethylbenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromo-5-(tert-butyl)-1,3-dimethylbenzene

B5924-1G B5924-5G
TCB5924-1GEA 100 USD
TCB5924-1G TCB5924-5G
2-Bromo-5-tert-butyl-1,3-dimethylbenzene ≥98.0% (by GC)
2-Bromo-5-tert-butyl-1,3-dimethylbenzene
Formula: C₁₂H₁₇Br
MW: 241.17 g/mol
MDL Number: MFCD12973868
CAS Number: 5345-05-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR