2-Butylbenzo[d]isothiazol-3(2H)-one ≥95.0% (by GC)

Supplier: TCI America
Danger

B6021-100G B6021-25G
TCB6021-100GEA 268 USD
TCB6021-100G TCB6021-25G
2-Butylbenzo[d]isothiazol-3(2H)-one ≥95.0% (by GC)
2-Butylbenzo[d]isothiazol-3(2H)-one
Formula: C₁₁H₁₃NOS
Boiling Pt: 180 °C
MDL Number: MFCD09751282
CAS Number: 4299-07-4
EINECS: 420-590-7
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR