(Chloromethyl)dimethoxy(methyl)silane ≥97.0% (by GC)

Supplier: TCI America
Warning

C3256-1G C3256-5G
TCC3256-1GEA 31 USD
TCC3256-1G TCC3256-5G
(Chloromethyl)dimethoxy(methyl)silane ≥97.0% (by GC)
(Chloromethyl)dimethoxy(methyl)silane
Formula: C₄H₁₁ClO₂Si
Boiling Pt: 140 °C
Flash Pt: 55 °C
MDL Number: MFCD04974117
CAS Number: 2212-11-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR