(Chloromethyl)diethoxy(methyl)silane ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Chloromethylmethyldiethoxysilane

C3257-25G C3257-5G
TCC3257-25GEA 167 USD
TCC3257-25G TCC3257-5G
(Chloromethyl)diethoxy(methyl)silane ≥98.0% (by GC)
(Chloromethyl)diethoxy(methyl)silane
Formula: C₆H₁₅ClO₂Si
MW: 182.72 g/mol
Boiling Pt: 161 °C
Flash Pt: 37 °C
MDL Number: MFCD00018878
CAS Number: 2212-10-4
EINECS: 218-657-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR