BX-471 ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (R)-1-[5-Chloro-2-[2-[4-(4-fluorobenzyl)-2-methylpiperazin-1-yl]-2-oxoethoxy]phenyl]urea, BX471, (R)-1-(5-Chloro-2-(2-(4-(4-fluorobenzyl)-2-methylpiperazin-1-yl)-2-oxoethoxy)phenyl)urea

B6060-25MG B6060-5MG
TCB6060-25MGEA 502 USD
TCB6060-25MG TCB6060-5MG
BX-471 ≥98.0% (by HPLC)
BX-471
Formula: C₂₁H₂₄ClFN₄O₃
MW: 434.89 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09859709
CAS Number: 217645-70-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 11.60 to 13.50 %
Water (Vaporization methoed) max. 8.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR