ABT751 ≥98.0% (by HPLC)

Supplier: TCI America

A3169-10MG A3169-50MG
TCA3169-10MGEA 121 USD
TCA3169-10MG TCA3169-50MG
ABT751 ≥98.0% (by HPLC)
ABT751
Formula: C₁₈H₁₇N₃O₄S
MW: 371.41 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00910291
CAS Number: 141430-65-1

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 10.70 to 11.90 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR