Dihydrotanshinone I ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: DHT, (R)-1,6-Dimethyl-1,2-dihydrophenanthro[1,2-b]furan-10,11-dione, (1R)-1,6-dimethyl-1,2-dihydronaphtho[1,2-g][1]benzofuran-10,11-dione, (-)-Dihydrotanshinone I, 15,16-Dihydrotanshinone I

D5379-10MG D5379-50MG
TCD5379-10MGEA 89 USD
TCD5379-10MG TCD5379-50MG
Dihydrotanshinone I ≥97.0% (by HPLC)
Dihydrotanshinone I
Formula: C₁₈H₁₄O₃
MW: 278.3 g/mol
MDL Number: MFCD28016070
CAS Number: 87205-99-0

Order Now

Specification Test Results

Appearance Red to Dark red to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 214.0 to 218.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR