SCR7 pyrazine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2,3-Dihydro-6,7-diphenyl-2-thioxo-4(1H)-pteridinone, 6,7-Diphenyl-2-thioxo-2,3-dihydropteridin-4(1H)-one

C3577-10MG C3577-50MG
TCC3577-10MGEA 126 USD
TCC3577-10MG TCC3577-50MG
SCR7 pyrazine ≥98.0% (by HPLC)
SCR7 pyrazine
Formula: C₁₈H₁₂N₄OS
Storage Temperature: Refrigerator
MDL Number: MFCD02167478
CAS Number: 14892-97-8

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 207.0 to 211.0 deg-C
Elemental analysis (Nitrogen) 16.00 to 17.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR