SCH-58261 ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: SCH 58261, 2-(furan-2-yl)-7-phenethyl-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine, SCH58261, SCH-58261

C3599-25MG C3599-5MG
TCC3599-25MGEA 480 USD
TCC3599-25MG TCC3599-5MG
SCH-58261 ≥98.0% (by HPLC)
SCH-58261
Formula: C₁₈H₁₅N₇O
Storage Temperature: Refrigerator
MDL Number: MFCD00933778
CAS Number: 160098-96-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 221.0 to 225.0 deg-C
Elemental analysis (Nitrogen) 27.00 to 29.80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR