Afatinib ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

A2870-250MG A2870-50MG
TCA2870-250MGEA 187 USD
TCA2870-250MG TCA2870-50MG
Afatinib ≥97.0% (by HPLC, total nitrogen)
Afatinib
Formula: C₂₄H₂₅ClFN₅O₃
Melting Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12407405
CAS Number: 850140-72-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR