Cyclothiazide ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 6-Chloro-3,4-dihydro-3-(2-norbornen-5-yl)-2H-1,2-4-benzothiadiazine-7-sulphonamide 1,1-Dioxide, Cyclothiazide (mixture of isomers)

C3392-100MG C3392-25MG
TCC3392-100MGEA 248 USD
TCC3392-100MG TCC3392-25MG
Cyclothiazide ≥95.0% (by HPLC)
Cyclothiazide
Formula: C₁₄H₁₆ClN₃O₄S₂
Melting Pt: 234 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00210192
CAS Number: 2259-96-3
EINECS: 218-859-7
Merck Index: 14,02754

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 10.20 to 11.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR