SC-560 ≥98.0% (by HPLC)

Supplier: TCI America

C3572-100MG C3572-25MG
TCC3572-100MGEA 236 USD
TCC3572-100MG TCC3572-25MG
SC-560 ≥98.0% (by HPLC)
SC-560
Formula: C₁₇H₁₂N₂OClF₃
Storage Temperature: Refrigerator
MDL Number: MFCD02179214
CAS Number: 188817-13-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 7.50 to 8.50 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR