Cambinol ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: CAmbinol, 5-[(2-Hydroxy-1-naphthyl)methyl]-6-phenyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone, 5-[(2-Hydroxynaphthalen-1-yl)methyl]-6-phenyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one, NSC 112546

C3535-25MG C3535-5MG
TCC3535-25MGEA 451 USD
TCC3535-25MG TCC3535-5MG
Cambinol ≥98.0% (by HPLC)
Cambinol
Formula: C₂₁H₁₆N₂O₂S
MW: 360.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09057543
CAS Number: 14513-15-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR