1,4-Cineole ≥93.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Isopropyl-4-methyl-7-oxabicyclo[2.2.1]heptane

C3652-100G C3652-25G
TCC3652-100GEA 99 USD
TCC3652-100G TCC3652-25G
1,4-Cineole ≥93.0% (by GC)
1,4-Cineole
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 173 °C
Melting Pt: -46 °C
Flash Pt: 47 °C
MDL Number: MFCD00209502
CAS Number: 470-67-7
EINECS: 207-428-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR