3-Aminomethyltetrahydrofuran ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (Tetrahydrofuran-3-yl)methanamine, 3-(Aminomethyl)oxolane, (tetrahydrofuran-3-yl)methanamine, Oxolan-3-ylmethanamine, 3-(Aminomethyl)tetrahydrofuran

A3239-1G A3239-5G
TCA3239-1GEA 74 USD
TCA3239-1G TCA3239-5G
3-Aminomethyltetrahydrofuran ≥97.0% (by GC)
3-Aminomethyltetrahydrofuran
Formula: C₅H₁₁NO
Boiling Pt: 67 °C
Flash Pt: 58 °C
MDL Number: MFCD08234925
CAS Number: 165253-31-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR